Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPGDS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103857 Supplier Invitrogen™ Supplier No. PA5103857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PTGDS2.

PGDS is a sigma class glutathione-S-transferase family member. The enzyme catalyzes the conversion of PGH2 to PGD2 and plays a role in the production of prostanoids in the immune system and mast cells. The presence of this enzyme can be used to identify the differentiation stage of human megakaryocytes. Prostaglandin-D synthase is a sigma class glutathione-S-transferase family member. The enzyme catalyzes the conversion of PGH2 to PGD2 and plays a role in the production of prostanoids in the immune system and mast cells. The presence of this enzyme can be used to identify the differentiation stage of human megakaryocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPGDS
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Hpgds
Gene Accession No. O60760
Gene Alias EC 2.5.1.18; EC 5.3.99.2; glutathione S-transferase; glutathione S-transferase sigma; glutathione-dependent PGD synthase; glutathione-dependent PGD synthetase; glutathione-requiring prostaglandin D synthase; GST class-sigma; GSTS; GSTS1; GSTS1-1; hematopoietic prostaglandin D synthase; hematopoietic prostaglandin D2 synthase; Hpgds; H-PGDS; PGD2; PGDS; prostaglandin D2 synthase 2, hematopoietic; prostaglandin D2 synthase, hematopoietic; prostaglandin-H2 D-isomerase; Ptgds2
Gene Symbols Hpgds
Host Species Rabbit
Immunogen A synthesized peptide derived from human HPGDS(Accession O60760), corresponding to amino acid residues L54-W104.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27306
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.