Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPi2 Antibody (HIC1-2B4.2B), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP118946PECY55
Description
HPi2 Monoclonal specifically detects HPi2 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
HPi2 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Tissue culture supernatant | |
Primary | |
Human, Mouse | |
Supernatant |
Flow Cytometry | |
HIC1-2B4.2B | |
Flow Cytometry | |
human pancreatic endocrine islet cell | |
Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
0.1 mL | |
Specific for multiple endocrine cell types. | |
Store at 4°C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction