Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPi3 Antibody (HIC1-7H10), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP118947APCCY7

Catalog No. N118947APC7


Only null left
Add to Cart

Description

Description

HPi3 Monoclonal specifically detects HPi3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

HPi3
Monoclonal
APC-Cyanine7
PBS with 0.05% Sodium Azide
Mouse
Tissue culture supernatant
Primary
Human, Mouse
Supernatant
Flow Cytometry
HIC1-7H10
Flow Cytometry
human pancreatic endocrine islet cell
Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells.
0.1 mL
Specific for multiple endocrine cell types.
Store at 4°C in the dark. Do not freeze.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.