Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPi4 Antibody (HIC1-5F10), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP118948MFV450
Description
HPi4 Monoclonal antibody specifically detects HPi4 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
HPi4 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Tissue culture supernatant | |
RUO | |
Human, Mouse | |
Supernatant |
Flow Cytometry | |
HIC1-5F10 | |
Flow Cytometry | |
human pancreatic endocrine islet cell | |
Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction