Learn More
HPIP Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301629A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Regulator of pre-B-cell leukemia transcription factors (BPXs) function. Inhibits the binding of PBX1-HOX complex to DNA and blocks the transcriptional activity of E2A-PBX1. Tethers estrogen receptor-alpha (ESR1) to microtubules and allows them to influence estrogen receptors-alpha signaling.Specifications
| HPIP | |
| Polyclonal | |
| Unconjugated | |
| PBXIP1 | |
| pre-B-cell leukemia homeobox interacting protein 1, HPIP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57326 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96AQ6 | |
| PBXIP1 | |
| Between 500 and 550 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.