Learn More
Promega HPK1 Kinase Enzyme System, 1mg
Catalog No. PRV6398 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6398 1 mg
1 options
Catalog No. PRV6398 Supplier Promega Supplier No. V6398
Add to Cart
Add to Cart
Request a Quote
Recombinant human HPK1 (1-346) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HPK1 or Hematopoietic progenitor kinase 1 is a hematopoietic cell-restricted member of the Ste20 serine/threonine kinase super family.
  • Easily Screen and Profile HPK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C