Learn More
HPK1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301918AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May play a role in the response to environmental stress. Appears to act upstream of the JUN N-terminal pathway. May play a role in hematopoietic lineage decisions and growth regulation.Specifications
HPK1 | |
Polyclonal | |
Unconjugated | |
MAP4K1 | |
MAP4K1; mitogen-activated protein kinase kinase kinase kinase 1; HPK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11184 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q92918 | |
MAP4K1 | |
Between 550 and 600 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.