Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPRT Antibody (2G8), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174045AF350
Description
HPRT Monoclonal specifically detects HPRT in Human samples. It is validated for Western Blot, ELISA.Specifications
HPRT | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
HPRT1 | |
Recombinant human HPRT (1-218aa) purified from E. coli | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA | |
2G8 | |
Western Blot, ELISA | |
EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
Mouse | |
100 μL | |
3251 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction