Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPRT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$489.50
Specifications
| Antigen | HPRT |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
HPRT Polyclonal specifically detects HPRT in Human, Mouse samples. It is validated for Western Blot.Specifications
| HPRT | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
| HPRT1 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P00492 | |
| 3251 | |
| IgG | |
| 24 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title