Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPRT Antibody (OTI2F7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270934F
Description
HPRT Monoclonal specifically detects HPRT in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
HPRT | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
HPRT1 | |
Full length recombinant protein of human HPRT (NP_000185) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunocytochemistry, Immunofluorescence | |
OTI2F7 | |
Immunocytochemistry/Immunofluorescence | |
EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
3251.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction