Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPRT Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30336120UL
Description
HPRT Polyclonal specifically detects HPRT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
HPRT | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence, KnockDown | |
Unconjugated | |
EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-218 of human HPRT (NP_000185.1). | |
20 μL | |
Lipid and Metabolism | |
3251 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction