Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPRT1 Monoclonal Antibody (571CT1.3.4)
GREENER_CHOICE

Catalog No. PIMA537588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537588 Supplier Invitrogen™ Supplier No. MA537588
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPRT1 Monoclonal Antibody for Western Blot

The protein is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPRT1
Applications Western Blot
Classification Monoclonal
Clone 571CT1.3.4
Concentration Conc. not determined
Conjugate Unconjugated
Formulation ascites with 0.09% sodium azide; pH 7.4
Gene HPRT1
Gene Accession No. P00492
Gene Alias C81579; HGPRT; HGPRTase; HPGRT; Hprt; HPRT 1; HPRT B; Hprt1; hprt1 protein; hypoxanthine guanine phosphoribosyl transferase; hypoxanthine guanine phosphoribosyl transferase 1; hypoxanthine phosphoribosyl transferase; hypoxanthine phosphoribosyltransferase 1; hypoxanthine-guanine phosphoribosyltransferase; hypoxanthine-guanine phosphoribosyltransferase 1; id:ibd1344; id:ibd5108; testicular tissue protein Li 89; wu:fc10g09; zgc:56221; zgc:86608
Gene Symbols HPRT1
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 150-178 amino acids from human HPRT1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3251
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.