Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPS1 Monoclonal Antibody (5G12G2)
GREENER_CHOICE

Catalog No. PIMA515381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515381 Supplier Invitrogen™ Supplier No. MA515381
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15381 targets HPS1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15381 immunogen is purified recombinant fragment of HPS1 expressed in E. Coli..

Hermansky-Pudlak syndrome (HPS) is a rare, genetically heterogeneous, autosomal recessive disorder. It is characterized by oculocutaneous albinism, lysosomal storage defects and prolonged bleeding due to platelet storage pool deficiency. There are 10 HPS genes encoding HPS proteins that all interact within three distinct ubiquitously expressed protein complexes or biogenesis of lysosome-related organelle complexes. Defects in these genes cause HPS. HSP-1 is a component of multiple cytoplasmic organelles and is important for their normal development and function. It plays a role in intracellular protein sorting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPS1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5G12G2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene HPS1
Gene Accession No. Q92902
Gene Alias 6030422N11Rik; BB405864; BLOC3S1; ep; Gm21361; Hermansky-Pudlak syndrome 1 homolog (human); Hermansky-Pudlak syndrome 1 protein; Hermansky-Pudlak syndrome 1 protein homolog; Hermansky-Pudlak syndrome protein homolog; HPS; HPS1; HPS1, biogenesis of lysosomal organelles complex 3 subunit 1; MGC5277; pale ear
Gene Symbols HPS1
Host Species Mouse
Immunogen Purified recombinant fragment of HPS1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.