Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563938
Description
Immunogen sequence: LWQGINLVLL TRSPSAPLAL VLSQLMDGFS MLEKKLKEGP EPGASLRSQP LVGDLRQRMD KFVKNRGAQE IQSTWLEFKA KAFSK Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.
Hermansky-Pudlak syndrome (HPS) is a rare, genetically heterogeneous, autosomal recessive disorder. It is characterized by oculocutaneous albinism, lysosomal storage defects and prolonged bleeding due to platelet storage pool deficiency. There are 10 HPS genes encoding HPS proteins that all interact within three distinct ubiquitously expressed protein complexes or biogenesis of lysosome-related organelle complexes. Defects in these genes cause HPS. HSP-1 is a component of multiple cytoplasmic organelles and is important for their normal development and function. It plays a role in intracellular protein sorting.
Specifications
HPS1 | |
Polyclonal | |
Unconjugated | |
HPS1 | |
6030422N11Rik; BB405864; BLOC3S1; ep; Gm21361; Hermansky-Pudlak syndrome 1 homolog (human); Hermansky-Pudlak syndrome 1 protein; Hermansky-Pudlak syndrome 1 protein homolog; Hermansky-Pudlak syndrome protein homolog; HPS; HPS1; HPS1, biogenesis of lysosomal organelles complex 3 subunit 1; MGC5277; pale ear | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3257 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92902 | |
HPS1 | |
Recombinant protein corresponding to Human HPS1. Recombinant protein control fragment (Product #RP-101171). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction