Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111362
Description
Immunogen sequence: YVNWRNKRTE NSRVCIRMIG HNVEGPFSKA FRDQMYIIEM PLSEAPLCIS CCPVKGDLLV GCTNKLVLFS LKYQIINEEF SLLDFER.
This gene encodes a protein containing a potential clathrin-binding motif, consensus dileucine signals, and tyrosine-based sorting signals for targeting to vesicles of lysosomal lineage. The encoded protein may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 3. Alternate splice variants exist, but their full length sequence has not been determined.
Specifications
| HPS3 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969F9 | |
| Hps3 | |
| Recombinant Protein corresonding to Human HPS3. Recombinant protein control fragment (Product #RP-106416). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| Hps3 | |
| BLOC2S1; coa; cocoa; Cocoa protein; DKFZp686F0413; FLJ22704; Hermansky-Pudlak syndrome 3; Hermansky-Pudlak syndrome 3 homolog (human); Hermansky-Pudlak syndrome 3 protein; Hermansky-Pudlak syndrome 3 protein homolog; hps3; HPS3 biogenesis of lysosomal organelles complex 2 subunit 1; HPS3, biogenesis of lysosomal organelles complex 2 subunit 1; SUTAL; zgc:152969 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84343 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction