Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115380 Supplier Invitrogen™ Supplier No. PA5115380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HPS3.

This gene encodes a protein containing a potential clathrin-binding motif, consensus dileucine signals, and tyrosine-based sorting signals for targeting to vesicles of lysosomal lineage. The encoded protein may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 3. Alternate splice variants exist, but their full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPS3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Hps3
Gene Accession No. Q91VB4, Q969F9
Gene Alias BLOC2S1; coa; cocoa; Cocoa protein; DKFZp686F0413; FLJ22704; Hermansky-Pudlak syndrome 3; Hermansky-Pudlak syndrome 3 homolog (human); Hermansky-Pudlak syndrome 3 protein; Hermansky-Pudlak syndrome 3 protein homolog; hps3; HPS3 biogenesis of lysosomal organelles complex 2 subunit 1; HPS3, biogenesis of lysosomal organelles complex 2 subunit 1; SUTAL; zgc:152969
Gene Symbols Hps3
Host Species Rabbit
Immunogen A synthesized peptide derived from human HPS3(Accession Q969F9), corresponding to amino acid residues S537-V587.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12807, 310288, 84343
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.