Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HPS3 Full-length ORF (AAH16901.1, 1 a.a. - 372 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947854 Shop All Abnova Corporation Products
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein containing a potential clathrin-binding motif, consensus dileucine signals, and tyrosine-based sorting signals for targeting to vesicles of lysosomal lineage. The encoded protein may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 3. Alternate splice variants exist, but their full length sequence has not been determined. [provided by RefSeq]

Sequence: MFYVAEPKQVPHILCSPSMKNINPLTAMSYLRKLDTSGFSSILVTLTKAAVALKMGDLDMHRNEMKSHSEMKLVCGFILEPRLLIQQRKGQIVPTELALHLKETQPGLLVASVLGLQKNNKIGIEEADSFFKVLCAKDEDTIPQLLVDFWEAQLVACLPDVVLQELFFKLTSQYIWRLSKRQPPDTTPLRTSEDLINACSHYGLIYPWVHVVISSDSLADKNYTEDLSKLQSLICGPSFDIASIIPFLEPLSEDTIAGLSVHVLCRTRLKEYEQCIDILLERCPEAVIPYANHELKEENRTLWWKKLLPELCQRIKCGGEKYQLYLSSLKETLSIVAVELELKDFMNVLPEDGTATFFLPYLLYCSRKKPLT

Specifications

Accession Number AAH16901.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84343
Molecular Weight (g/mol) 68.7kDa
Name HPS3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MFYVAEPKQVPHILCSPSMKNINPLTAMSYLRKLDTSGFSSILVTLTKAAVALKMGDLDMHRNEMKSHSEMKLVCGFILEPRLLIQQRKGQIVPTELALHLKETQPGLLVASVLGLQKNNKIGIEEADSFFKVLCAKDEDTIPQLLVDFWEAQLVACLPDVVLQELFFKLTSQYIWRLSKRQPPDTTPLRTSEDLINACSHYGLIYPWVHVVISSDSLADKNYTEDLSKLQSLICGPSFDIASIIPFLEPLSEDTIAGLSVHVLCRTRLKEYEQCIDILLERCPEAVIPYANHELKEENRTLWWKKLLPELCQRIKCGGEKYQLYLSSLKETLSIVAVELELKDFMNVLPEDGTATFFLPYLLYCSRKKPLT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686F0413/FLJ22704/SUTAL
Common Name HPS3
Gene Symbol HPS3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.