Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPS4 Polyclonal Antibody

Rabbit Polyclonal Antibody

$237.00 - $577.00

Specifications

Antigen HPS4
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119189
View Documents
Invitrogen™
PA5119189
200 μL
Each for $577.00
Only null left
Add to Cart
 
PIPA5121899
View Documents
Invitrogen™
PA5121899
20 μL
Each for $237.00
Only null left
Add to Cart
 
Description

Description

HPS4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
Specifications

Specifications

HPS4
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Mouse
HPS4
2010205O06Rik; AU040608; BLOC-3; BLOC3S2; C130020P05Rik; Hermansky-Pudlak syndrome 4; Hermansky-Pudlak syndrome 4 homolog; Hermansky-Pudlak syndrome 4 homolog (human); Hermansky-Pudlak syndrome 4 protein; Hermansky-Pudlak syndrome 4 protein homolog; Hps4; HPS4, biogenesis of lysosomal organelles complex 3 subunit 2; KIAA1667; Le; Le protein; light ear; light-ear protein; Light-ear protein homolog; mKIAA1667
HPS4
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q99KG7
192232
Recombinant protein Hermansky Pudlak Syndrome Protein 4. The antigen corresponds to amino acid range 20-214 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.