Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPS4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562152
Description
Immunogen sequence: GQRGNKLPTG EQGLDEDVDG VCESHAAPGL ECSSGSANCQ GAGPSADGIS SRLTPAESCM GLVRMNLYTH CVKGLVLSLL AEEPLLGDSA AIEEVYHSSL ASLNGLEVHL KETLPRDEAA STSSTYNFTH YDR Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 58%.
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
Specifications
HPS4 | |
Polyclonal | |
Unconjugated | |
HPS4 | |
2010205O06Rik; AU040608; BLOC-3; BLOC3S2; C130020P05Rik; Hermansky-Pudlak syndrome 4; Hermansky-Pudlak syndrome 4 homolog; Hermansky-Pudlak syndrome 4 homolog (human); Hermansky-Pudlak syndrome 4 protein; Hermansky-Pudlak syndrome 4 protein homolog; Hps4; HPS4, biogenesis of lysosomal organelles complex 3 subunit 2; KIAA1667; Le; Le protein; light ear; light-ear protein; Light-ear protein homolog; mKIAA1667 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89781 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQG7 | |
HPS4 | |
Recombinant protein corresponding to Human HPS4. Recombinant protein control fragment (Product #RP-91492). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction