Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595699 Supplier Invitrogen™ Supplier No. PA595699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAFLHVPVPD GRAPYCKASL SASSSLEPTP PEDTAISSLR PPSAPEMLTQ HGAQEQLEDH PGHSSQAPIP RADPLPRRTR RPLLLPRLDP GQRGNKLPTG EQGLDEDVDG VCESHAAPGL ECSSGSANCQ GAGPSADGIS SRLTPAESCM GLVRMNLYTH CVKGLVLSLL AEEPLLGDSA AIEEVYHSSL ASLNGLEVHL KETLPRDEAA STSSTYNFTH YDRIQSLLMA NLPQVATPQD RRFLQAVSLM HSEFAQLPAL YEMTVRNAST AVYACCNPIQ ETYFQQLAPA ARSSGFPNPQ DGAFSLSGKA KQKLLKHGVN LL Positive Samples: U-87MG, SGC-7901, LO2, Jurkat, Mouse liver.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPS4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HPS4
Gene Accession No. Q99KG7, Q9NQG7
Gene Alias 2010205O06Rik; AU040608; BLOC-3; BLOC3S2; C130020P05Rik; Hermansky-Pudlak syndrome 4; Hermansky-Pudlak syndrome 4 homolog; Hermansky-Pudlak syndrome 4 homolog (human); Hermansky-Pudlak syndrome 4 protein; Hermansky-Pudlak syndrome 4 protein homolog; Hps4; HPS4, biogenesis of lysosomal organelles complex 3 subunit 2; KIAA1667; Le; Le protein; light ear; light-ear protein; Light-ear protein homolog; mKIAA1667
Gene Symbols HPS4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 387-708 of human HPS4 (NP_0713644).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192232, 304555, 89781
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.