Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPS5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549270
Description
HPS5 Polyclonal Antibody for Western Blot
May regulate the synthesis and function of lysosomes and of highly specialized organelles, such as melanosomes and platelet dense granules. Regulates intracellular vesicular trafficking in fibroblasts. May be involved in the regulation of general functions of integrins.
Specifications
HPS5 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9UPZ3 | |
Hps5 | |
KLH conjugated synthetic peptide between 88-116 amino acids from the N-terminal region of human HPS5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Hps5 | |
Aa2-028; Ab1-018; AI646796; AIBP63; AL022647; alpha integrin binding protein 63; alpha-integrin-binding protein 63; BLOC2S2; C85120; haze; Hermansky-Pudlak syndrome 5; Hermansky-Pudlak syndrome 5 homolog (human); Hermansky-Pudlak syndrome 5 protein; Hermansky-Pudlak syndrome 5 protein homolog; HPS5; HPS5, biogenesis of lysosomal organelles complex 2 subunit 2; hz; KIAA1017; maroon; mr; Ru2; ru-2; ruby eye 2; ruby-eye protein 2; Ruby-eye protein 2 homolog | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
11234 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction