Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPS5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588334 Supplier Invitrogen™ Supplier No. PA588334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IISSRRGSQS DEDSCSLHSQ TLSEDERFKE FTSQQEEDLP DQCCGSHGNE DNVSHAPVMF ETDKNETFLP FGIPLPFRSP SPLVSLQAVK ESVSSFVRKT TEKIGTLHTS PDLKVRPELR GDEQSCEEDV SSDTCPKEED TEEEKEVTSP PPEEDRFQEL KVATAEAMTK LQDPLVLFES ESLRMVLQEW LSHLEKTFAM KDFSGVSDTD NSSMKLNQDV LLVNESKKGI LDEDNEKEKR DSLGNEESVD KTACECVRSP R; Positive Samples: Jurkat, 293T, B cells; Cellular Location: Cytoplasm, cytosol.

May regulate the synthesis and function of lysosomes and of highly specialized organelles, such as melanosomes and platelet dense granules. Regulates intracellular vesicular trafficking in fibroblasts. May be involved in the regulation of general functions of integrins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPS5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Hps5
Gene Accession No. Q9UPZ3
Gene Alias Aa2-028; Ab1-018; AI646796; AIBP63; AL022647; alpha integrin binding protein 63; alpha-integrin-binding protein 63; BLOC2S2; C85120; haze; Hermansky-Pudlak syndrome 5; Hermansky-Pudlak syndrome 5 homolog (human); Hermansky-Pudlak syndrome 5 protein; Hermansky-Pudlak syndrome 5 protein homolog; HPS5; HPS5, biogenesis of lysosomal organelles complex 2 subunit 2; hz; KIAA1017; maroon; mr; Ru2; ru-2; ruby eye 2; ruby-eye protein 2; Ruby-eye protein 2 homolog
Gene Symbols Hps5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 340-600 of human HPS5 (NP_0091473).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11234
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.