Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPS5 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS17383R

Description
May regulate the synthesis and function of lysosomes and of highly specialized organelles, such as melanosomes and platelet dense granules. Regulates intracellular vesicular trafficking in fibroblasts. May be involved in the regulation of general functions of integrins.Specifications
HPS5 | |
Polyclonal | |
Unconjugated | |
Hps5 | |
Aa2-028; Ab1-018; AI646796; AIBP63; AL022647; alpha integrin binding protein 63; Alpha-integrin-binding protein 63; BLOC2S2; C85120; haze; Hermansky-Pudlak syndrome 5; Hermansky-Pudlak syndrome 5 homolog (human); Hermansky-Pudlak syndrome 5 protein; Hermansky-Pudlak syndrome 5 protein homolog; Hps5; HPS5, biogenesis of lysosomal organelles complex 2 subunit 2; hz; KIAA1017; maroon; mr; Ru2; ru-2; ruby eye 2; Ruby-eye protein 2; Ruby-eye protein 2 homolog | |
Rabbit | |
Protein A | |
RUO | |
11234 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9UPZ3 | |
Hps5 | |
KLH conjugated synthetic peptide derived from human HPS5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction