Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518338
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518338 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518338 Supplier Invitrogen™ Supplier No. PA518338
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. This protein interacts with Hermansky-Pudlak syndrome 5 protein. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPS6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Hps6
Gene Accession No. Q86YV9
Gene Alias 5330434M19Rik; BLOC2; BLOC2S3; FLJ22501; Hermansky-Pudlak syndrome 6; Hermansky-Pudlak syndrome 6 protein; Hermansky-Pudlak syndrome 6 protein homolog; Hermansky-Pudlak syndrome-6 protein (HPS6); Hps6; HPS6, biogenesis of lysosomal organelles complex 2 subunit 3; Hsp6; MGC20522; RP11-302K17.1; ru; ruby eye; ruby-eye protein; Ruby-eye protein homolog; ruby-eye-like protein
Gene Symbols Hps6
Host Species Goat
Immunogen Synthetic peptide sequence (LRTELRDYRGLEC) corresponding to the internal amino acids of HPS6 (aa 455-465).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79803
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.