Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583510 Supplier Invitrogen™ Supplier No. PA583510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRGNLRLLSA LGLFCVGWEA PQGVELPSAK DLVFEEACGY YQRRSLRGAQ LTPEELRHSS TFRAPQALAS ILQGHLPPSA LLTM Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 89%.

This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. This protein interacts with Hermansky-Pudlak syndrome 5 protein. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPS6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hps6
Gene Accession No. Q86YV9
Gene Alias 5330434M19Rik; BLOC2; BLOC2S3; FLJ22501; Hermansky-Pudlak syndrome 6; Hermansky-Pudlak syndrome 6 protein; Hermansky-Pudlak syndrome 6 protein homolog; Hermansky-Pudlak syndrome-6 protein (HPS6); Hps6; HPS6, biogenesis of lysosomal organelles complex 2 subunit 3; Hsp6; MGC20522; RP11-302K17.1; ru; ruby eye; ruby-eye protein; Ruby-eye protein homolog; ruby-eye-like protein
Gene Symbols Hps6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HPS6. Recombinant protein control fragment (Product #RP-97152).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79803
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.