Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV 16 Monoclonal Antibody (ABM257), Abbomax

Catalog No. 502203767
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3767 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3767 Supplier Abbomax Inc Supplier No. 5005834
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: HPV infected tissue Cellular location: Nuclear.

HPV 16 Monoclonal antibody specifically detects HPV 16 in Virus samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed)

Specifications

Antigen HPV 16
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone ABM257
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias HPV L1; HPV16; HPV-16; human papillomavirus
Host Species Mouse
Immunogen A recombinant protein of HPV type 16 protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.