Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV L2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988637
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8637 Supplier Bioss Supplier No. BS8547R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Minor protein of the capsid that localizes along the inner surface of the virion, within the central cavities beneath the L1 pentamers. Plays a role in capsid stabilization through interaction with the major capsid protein L1. Once the virion enters the host cell, L2 escorts the genomic DNA into the nucleus by promoting escape from the endosomal compartments and traffic through the host Golgi network. Plays a role through its interaction with host dynein in the intracellular microtubule-dependent transport of viral capsid toward the nucleus. Mediates the viral genome import into the nucleus through binding to host importins. Once within the nucleus, L2 localizes viral genomes to host PML bodies in order to activate early gene expression for establishment of infection. Later on, promotes late gene expression by interacting with the viral E2 protein and by inhibiting its transcriptional activation functions. During virion assembly, encapsidates the genome by direct interaction with the viral DNA.

Specifications

Antigen HPV L2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from HPV16 L2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.