Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPV pan-specific L1 Monoclonal Antibody (5308)
GREENER_CHOICE

Catalog No. PIMA533267
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533267 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533267 Supplier Invitrogen™ Supplier No. MA533267
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reactive with the L1 of types 6, 11, 16, 18, 31, 33, 45, 52, & 58.

The papillomavirus major capsid protein, L1, is a ~55 kD protein with the ability to spontaneously self-assemble into virus-like particles (VLPs). These VLPs present an exterior surface essentially indistinguishable from the native 60 nm non-enveloped papillomavirus virion. Purified recombinant L1 proteins can achieve this complex assembly reaction in the absence of any chaperones (Schiller and Lowy, 2012). PMID: 23800545.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV pan-specific L1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5308
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Human Papillomavirus; Human Papillomavirus L1; Human Papillomavirus L1 Pan
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.