Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPV pan-specific L1 Monoclonal Antibody (5307)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA533266

Catalog No. PIMA533266

  • $630.00
    $508.65 / Each of 1

    Save $121.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart

Description

Description

Reactive with the L1 of types 6, 11, 16, 18, 31, 33, 45, 52, & 58.

The papillomavirus major capsid protein, L1, is a ~55 kD protein with the ability to spontaneously self-assemble into virus-like particles (VLPs). These VLPs present an exterior surface essentially indistinguishable from the native 60 nm non-enveloped papillomavirus virion. Purified recombinant L1 proteins can achieve this complex assembly reaction in the absence of any chaperones (Schiller and Lowy, 2012). PMID: 23800545.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HPV pan-specific L1
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
1 mL
Primary
4°C
Liquid
ELISA, Immunohistochemistry, Western Blot
5307
Unconjugated
Human Papillomavirus; Human Papillomavirus L1; Human Papillomavirus L1 Pan
Protein A
RUO
Virus
Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.