Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPV Type 16/18 E6 Monoclonal Antibody (C1P5)
GREENER_CHOICE

Catalog No. MA146057
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA146057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA146057 Supplier Invitrogen™ Supplier No. MA146057
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV Type 16/18 E6 Monoclonal Antibody for Western Blot, IHC (P), IP

Human Papilloma virus (HPV) is responsible for the development of type and tissue specific papilloma in the oral cavity and in the larynx. In the skin papilloma virus can cause different types of warts and in the ano-genital region it is associated with condyloma or carcinoma. HPV types 1, 6 and 11 usually are associated with benign transformations of the tissues, while e.g. HPV types 16, 18 and 31 seem to be responsible for the development of some carcinoma like cervical carcinoma. The role of HPV in non-melanoic tumours of the skin is under discussion. Protein E6 is a Transcriptional transactivator, binding double stranded DNA. It has transforming activity inactivating, with E6-AP ubiquitin-protein ligase, the human TP53/p53 tumor suppressor protein by targeting it for degradation. Clinical relevance: Detection of HPV in cervical smears and biopsies. Uses: Analysis of E6 expression in cell transformation studies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV Type 16/18 E6
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C1P5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Alias HPV16; HPV-16; HPV18; HPV-18; human papillomavirus
Host Species Mouse
Immunogen Gel-purified HPV-18 E6-beta galactosidase fusion protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.