Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPV Type 16 E7 Monoclonal Antibody (8C9)
GREENER_CHOICE

Catalog No. 280006
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-000-6 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-000-6 Supplier Invitrogen™ Supplier No. 280006
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

In Western blot against the HPV16-E7 bacterially derived fusion protein, the antibody detects a 52 kD band which is composed of the HPV16-E7 protein (15 kDa) and the trpE protein (37 kDa). Positive reactivity for the 15 kDa HPV16-E7 expressed protein was shown with CaSki (HPV16 DNA-containing cell line) cells by Western blot. The antibody has also been shown to immunoprecipitate and immunocytochemically stain the HPV16-E7 expressed protein from the CaSki cell line. Negative reactivity by Western blot was shown with a series of other HPV fusion proteins including: HPV16-E2, HPV16-E4, HPV16-E6, HPV18-E7, HPV6-E7, HPV6-E4, and HPV6-L1. The antibody failed to react with HeLa (HPV18 DNA-containing cell line) cells and other HPV DNA negative cervical carcinoma cell lines, HT-3 and C-33A, by Western blot, by immunoprecipitation or by immunocytochemical staining analyses. Reconstitute the lyophilized material with 1 mL of deionized or distilled water for a final concentration of 50 μg/mL.

The HPV E7 proteins are small (HPV16 E7 comprising 98 amino acids, 15kDa), zinc binding phosphoproteins which are localised in the nucleus. They are structurally and functionally similar to the E1A protein of subgenus C adenoviruses. The first 16 amino-terminal amino acids of HPV16 E7 contain a region homologous to a segment of the conserved region 1 (CR1) of the E1A protein of subgenus C adenoviruses. The next domain, up to amino acid 37, is homologous to the entire region 2 (CR2) of E1A. Genetic studies have established that these domains are required for cell transformation in vitro, suggesting similarities in the mechanism of transformation by these viruses. The CR2 homology region contains the LXCXE motif (residues 22-26) involved in binding to the tumour suppressor protein pRb.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV Type 16 E7
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8C9
Concentration 50 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide
Gene Alias HPV 16-E7; HPV16E7; human papillomavirus
Host Species Mouse
Immunogen Bacterially derived fusion protein containing the human papillomavirus tp 16 early protein E7 open reading frame.
Purification Method HPLC
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.