Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPV Type 18 E7 Monoclonal Antibody (GT881)
GREENER_CHOICE

Catalog No. PIMA535923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535923 Supplier Invitrogen™ Supplier No. MA535923
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

HPV Type 18 E7 Monoclonal Antibody for Western Blot, ICC/IF, ELISA, LF

Infection with specific types of HPV has been associated with an increased risk of developing cervical neoplasia. HPV types 6 and 11 have been associated with relatively benign diseases such as genital warts but types 16 and 18 are strongly associated with cervical, vaginal, and vulvar malignancies. E7-driven degradation of pRb may be involved in cervical tumorigenesis in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV Type 18 E7
Applications ELISA, Lateral Flow, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT881
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative
Gene Alias HPV E7; HPV18; HPV-18; human papillomavirus
Host Species Mouse
Immunogen Full length Human Papilloma virus type 18 E7 recombinant protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.