Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPV Type 18 L1 Monoclonal Antibody (5302)
GREENER_CHOICE

Catalog No. PIMA533262
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533262 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533262 Supplier Invitrogen™ Supplier No. MA533262
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV Type 18 L1 Monoclonal Antibody for ELISA

HPV Type 18 L1 is a protein that is a component of the human papillomavirus (HPV) Type 18 virus. This virus is one of the high-risk types of HPV that can cause cervical cancer and other types of cancer. The L1 protein is a major structural protein of the virus and is responsible for the formation of the viral capsid, which is the outer shell of the virus. The L1 protein is also a target for HPV vaccines, which can help prevent infection with HPV Type 18 and other high-risk types of HPV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV Type 18 L1
Applications ELISA
Classification Monoclonal
Clone 5302
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene L1
Gene Accession No. P06794
Gene Alias L1; Major capsid protein L1
Gene Symbols L1
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489090
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.