Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16 Antibody (CAMVIR-1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2452570.1MG
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Immunofluorescence
HPV16 Monoclonal specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
HPV16 | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
Human Papilloma Virus Type 16 | |
Human papilloma virus type 16, major capsid protein L1 | |
Protein A or G purified | |
RUO | |
Reacts with a protein of 57kDa, identified as the L1 protein of human papilloma virus type 16 (HPV-16). It is the major capsid protein of HPV-16. Infection with specific types of HPV has been associated with an increased risk of developing cervical neoplasia. HPV types 6 and 11 have been associated with relatively benign diseases such as genital warts but types 16 and 18 are strongly associated with cervical, vaginal, and vulvar malignancies. The antibody reacts very strongly with formalin-fixed, paraffin-embedded tissues containing HPV-16 or -33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV-6 or HPV-11. It cross-reacts with HPV37. | |
Store at 4C. | |
IgG2a κ |
Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
CAMVIR-1 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Mouse | |
57 kDa | |
0.1 mg | |
Primary | |
Human, Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction