Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16 Antibody (HPV16/2058R), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308554R
Description
HPV16 Monoclonal antibody specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry (Paraffin)Specifications
HPV16 | |
Monoclonal | |
DyLight 550 | |
Human Papilloma Virus Type 16 | |
Human HPV16, major capsid protein L1 | |
100 μg | |
Virology Bacteria and Parasites | |
Virus | |
Purified |
Immunohistochemistry (Paraffin) | |
HPV16/2058R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction