Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16 Antibody (SPM405), Alexa Fluor™ 350 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311575AF350
Description
HPV16 Monoclonal antibody specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
HPV16 | |
Monoclonal | |
Alexa Fluor 350 | |
Human Papilloma Virus Type 16 | |
Recombinant major capsid protein L1 from human papilloma virus type 16 | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2a κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SPM405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction