Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16 E1+E4 Mouse anti-Virus, Alexa Fluor 700, Clone: TVG 401, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250426AF700
Description
HPV16 E1+E4 Monoclonal specifically detects HPV16 E1+E4 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
HPV16 E1+E4 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry | |
TVG 401 | |
Western Blot, ELISA, Immunohistochemistry | |
ATP dependent helicase E1, E1, E4, HPV 16 ATP dependent helicase E1, HPV 16 E1, HPV 16 E4, HPV 16 replication protein E1, HPV16 ATP dependent helicase E1, HPV16 E1, HPV16 E4, HPV16 replication protein E1, Human papilloma virus type 16, Human papilloma virus type 16 ATP dependent helicase E1, Human papilloma virus type 16 E1, Human papilloma virus type 16 E4, Human papilloma virus type 16 replication protein E1, Human papillomavirus type 16, Human papillomavirus type 16 ATP dependent helicase E1, Human papillomavirus type 16 E1, Human papillomavirus type 16 E4, Human papillomavirus type 16 replication protein E1, Replication protein, Replication protein E1 | |
Antigen for hybridoma production was expressed as a b galactosidase fusion protein using the pEX expression system and was consequently cleaved to release the E1/\E4 polypeptide. | |
0.1 mL | |
Cancer, Virology Bacteria and Parasites | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction