Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 E2 Mouse anti-Virus, Clone: TVG 261, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen HPV16 E2
Clone TVG 261
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml
Applications Western Blot
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122514
SDP
View Documents
Novus Biologicals
NBP30752020UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

HPV16 E2 Monoclonal specifically detects HPV16 E2 in Virus samples. It is validated for Western Blot.
Specifications

Specifications

HPV16 E2
0.2 mg/ml
Western Blot
Unconjugated
Mouse
Virus
HPV16 regulatory protein E2, Human Papilloma Virus 16 E2, Human papilloma virus 16 regulatory protein E2, Human papillomavirus type 16 E2, Human papillomavirus type 16 regulatory protein E2, Regulatory protein E2
IgG1 κ
Protein A or G purified
TVG 261
Western Blot 1-2 ug/ml
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
Raised against Vaccinia-E2 followed by intravenous injection of the maltose binding protein MBP-E2
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.