Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16 E6 + HPV18 E6 Rabbit anti-Virus, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1719R

Specifications
HPV16 E6 + HPV18 E6 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
KLH conjugated synthetic peptides from HPV16 E6 + HPV18 E6. | |
100 μL | |
Primary | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction