Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16 E7 Antibody (716-325), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11017341F
Description
HPV16 E7 Monoclonal specifically detects HPV16 E7 in Virus samples. It is validated for ELISA, Sandwich ELISA.Specifications
HPV16 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
HPV oncoprotein E7, type 16 (both monomer and dimer forms according WB) | |
Store at 4C in the dark. | |
IgG2a |
ELISA | |
716-325 | |
ELISA, Sandwich ELISA | |
Human Papilloma Virus Type 16 | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 16. | |
0.2 mL | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction