Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16 E7 Antibody (716-332), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073151B200UL
Description
HPV16 E7 Monoclonal specifically detects HPV16 E7 in Virus samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
HPV16 E7 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
200 μL | |
Virus | |
Purified |
Western Blot, ELISA | |
716-332 | |
Western Blot, ELISA | |
E7, HPV16 E7, HpV16gp2, Links Transforming protein human papillomavirus 16, Protein E7, Transforming protein, Transforming protein human papillomavirus 16 | |
HPV oncoprotein E7, type 16. | |
HPV oncoprotein E7, type 16 (both monomer and dimer forms according WB) | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction