Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16/HPV18 E6 Antibody (HPV16/1295 + HPV18/1297), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308500IR
Description
HPV16/HPV18 E6 Monoclonal antibody specifically detects HPV16/HPV18 E6 in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
HPV16/HPV18 E6 | |
Monoclonal | |
DyLight 755 | |
E6, Human Papilloma Virus, Human papillomavirus type 16 E6, Human papillomavirus type 18 E6, Protein E6 | |
HPV18 E6-beta-galactosidase fusion protein | |
100 μg | |
Virology Bacteria and Parasites | |
Virus | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HPV16/1295 + HPV18/1297 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction