Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV18 E6 Antibody (C1N1) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200073952 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254807 0.1 mg
NB413811 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254807 Supplier Novus Biologicals Supplier No. NBP254807
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV18 E6 Monoclonal specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen HPV18 E6
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone C1N1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6
Host Species Mouse
Immunogen HPV18E6-beta-galactosidase fusion protein synthesized in E. coli
Molecular Weight of Antigen 16.5 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.