Learn More
Description
Specifications
Specifications
| Antigen | HPV18 E6 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HPV18/1297 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
