Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV18 E7 Antibody (718-15), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11017215AF405
Description
HPV18 E7 Monoclonal antibody specifically detects HPV18 E7 in Virus samples. It is validated for Western Blot, ELISA, KnockDown.Specifications
HPV18 E7 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
HPV oncoprotein E7, type 18. Cross-reacts with type 16. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, KnockDown | |
718-15 | |
Western Blot, ELISA, Sandwich ELISA, Knockdown Validated | |
E7 protein | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 18. | |
0.2 mL | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction