Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPx2 Antibody (HIC1-1C10), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 2 publications

$206.50 - $499.50

Specifications

Antigen HPx2
Clone HIC1-1C10
Concentration 1 mg/mL
Dilution Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP118952SS
SDP
View Documents
Novus Biologicals
NBP118952SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP118952
SDP
View Documents
Novus Biologicals
NBP118952
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

HPx2 Monoclonal specifically detects HPx2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

HPx2
1 mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Pancreatic Stem Cell Markers, Stem Cell Markers
Tissue culture supernatant with 0.05% Sodium Azide
Human pancreatic enriched exocrine cells
Primary
Store at 4C. Do not freeze.
HIC1-1C10
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100
Monoclonal
Supernatant
RUO
Human
human pancreatic exocrine cell
IgM
Tissue culture supernatant
Specific for exocrine cell types.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.