Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HR23AGoat anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501991562
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1562 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1562 Supplier Rockland Immunochemicals Supplier No. 600101291
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Reactivity occurs against human HR23A protein. Sequence homology as assessed by BLAST indicated 100% homology for this protein from human and rabbit. Cross reactivity with HR23A protein from dog, mouse and rat may also occur as sequence homology varies by one or two amino acid residues in this sequence by BLAST analysis. Reactivity with HR23A protein from other sources is not known.

The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.

Specifications

Antigen HR23A
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene RAD23A
Gene Accession No. P54725
Gene Alias 2310040P19Rik; AL024030; BOS_7437; HHR23A; HR23A; MGC111083; Mhr23a; RAD23 homolog A; RAD23 homolog A (S. cerevisiae); RAD23 homolog A, nucleotide excision repair protein; RAD23, yeast homolog, A; Rad23a; RAD23a homolog (S. cerevisiae); UV excision repair protein RAD23 homolog A
Gene Symbols RAD23A
Host Species Goat
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5886
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.