Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HR6A/UBE2A Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100554G
Description
HR6A/UBE2A Polyclonal antibody specifically detects HR6A/UBE2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
HR6A/UBE2A | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 6.3.2.19, hHR6A, HR6A, RAD6ARAD6 homolog A, UBC2, Ubiquitin carrier protein A, ubiquitin-conjugating enzyme E2 A, ubiquitin-conjugating enzyme E2A (RAD6 homolog), Ubiquitin-protein ligase A | |
A synthetic peptide which maps to a region between residues 10 and 50 of human ubiquitin-conjugating enzyme E2A using the numbering given in Swiss-Prot entry P49459 (GeneID 7319). | |
0.1 mL | |
DNA Repair, Ubiquitin Proteasome Pathway | |
7319 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction