Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HR6B/UBE2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220781
Description
HR6B/UBE2B Polyclonal antibody specifically detects HR6B/UBE2B in Human, Mouse samples. It is validated for Western Blot.Specifications
HR6B/UBE2B | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B | |
Rabbit | |
17 kDa | |
0.1 mL | |
Primary | |
Mouse. Species reactivity based on sequence identity: Human (100%), Rabbit (100%), Rat (100%), (95%), Bovine (100%), Porcine (100%), Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1.02mg/mL | |
Western Blot 1:500-1:3000 | |
P63146 | |
UBE2B | |
Recombinant protein encompassing a sequence within the center region of human UBE2B. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
7320 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction