Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HR6B/UBE2B Antibody (OTI1E11), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271337C
Description
HR6B/UBE2B Monoclonal specifically detects HR6B/UBE2B in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HR6B/UBE2B | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
UBE2B | |
Full length recombinant protein of human HR6B/UBE2B (NP_003328) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot | |
OTI1E11 | |
Western Blot | |
E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins | |
7320.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction